missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ resDNASEQ™ Quantitative CHO DNA Kits

This quantitative PCR (qPCR)-based kit, part of the resDNASEQ™ Quantitative CHO DNA System, allows users to detect sub-picogram amounts of residual DNA from CHO cell line in under 5 hours.

£3175.00

Specifications

Detection Method Primer-probe
Format Kit
PCR Method qPCR
Host Cell CHO
Label or Dye FAM
View More Specs

Products 1
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
30000682
View Documents
Applied Biosystems™
4402085
Assay Kit only
£3175.00
Each
Please to purchase this item. Need a web account? Register with us today!
Description
Show More
Specifications

Specifications

Primer-probe
qPCR
FAM
100 reactions
Kit
CHO
resDNASEQ™
100 Reactions
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Applied Biosystems™ resDNASEQ™ Quantitative CHO DNA Kits

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.