Learn More
Applied Biosystems™ resDNASEQ™ Quantitative E. coli DNA Kits
This quantitative PCR (qPCR)-based kit, part of the resDNASEQ™ E. coli Residual DNA Quantitation System, enables highly sensitive quantitation of host cell DNA from E. coli expression systems, typically in less than 4 hours.
£3730.00
Specifications
Detection Method | Primer-probe |
---|---|
Format | Kit |
PCR Method | qPCR |
Host Cell | E.coli |
Label or Dye | FAM |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
30000687
|
Applied Biosystems™
4460366 |
Assay Kit and Sample Preparation |
£3730.00
100 reactions |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This quantitative PCR (qPCR)-based kit, part of the resDNASEQ™ E. coli Residual DNA Quantitation System, enables highly sensitive quantitation of host cell DNA from E. coli expression systems, typically in less than 4 hours. The kit is part of an integrated system that includes sample preparation, TaqMan™ Assay and master mix, standard DNA, instrument, and software. The system overcomes the limitations of traditional methods by combining high-recovery PrepSEQ™ sample preparation and TaqMan™ Assay-based quantitation of residual DNA.
Features of the resDNASEQ Quantitative E. coli DNA Kit include:
• Highly sensitive quantitation using proven TaqMan™ real-time qPCR technology
• Manual and automated sample preparation, optimized for quantitative recovery from complex sample matrices (Table 1)
• Consistent performance across the expected range of DNA fragment sizes
• Integrated system from sample to results with sample preparation, master mix, TaqMan™ primer/probe mix, and genomic DNA standard
This kit is available with or without sample preparation reagents in the form of a PrepSEQ™ Residual DNA Sample Preparation Kit, which is optimized for highly efficient residual DNA recovery from complex mixtures of proteins, buffers, and salts.
Rapid, Sensitive Detection of E.Coli DNA
The resDNASEQ™ E. coli Residual DNA Quantitation System is a quantitative PCR (qPCR)-based system for the detection of host cell DNA from E. coli, an expression system commonly used for the production of recombinant proteins. Reliable and rapid, the system enables sensitive (LOQ = 15 pg DNA/mL test sample) and specific quantitation of E. coli DNA, typically in less than 4 hours. This performance helps ensure a high degree of confidence in quantitation data obtained from a broad range of sample types—from in-process samples to bulk drug substances—whether a sample contains high molecular weight or sheared DNA.
Table 1. DNA recovery using manual PrepSEQ™ sample preparation protocol.
Assay performance data using 10 pg E. coli genomic DNA spike per sample, 3 analysts, and 9 test samples.
50 mg/mL protein sample | ||
Mean Recovery | Mean %CV | |
E. coli | 83% | 5.04% |
Specifications
Primer-probe | |
qPCR | |
FAM | |
100 Reactions |
Kit | |
E.coli | |
resDNASEQ™ |
For Research Use Only. Not for use in diagnostic procedures.