Learn More
Applied Biosystems™ resDNASEQ™ Quantitative Vero DNA Kits
Shop All Applied Biosystems ProductsDescription
The resDNASEQ Quantitative Vero DNA Kit is a quantitative PCR (qPCR)-based system for the detection of host cell DNA from Vero (African Green Monkey) cells, an expression system commonly used for the production of vaccines. Reliable and rapid, the resDNASEQ system enables sensitive (as low as 1.5 pg DNA/mL test sample) and specific quantitation of Vero cell DNA, typically in less than four hours. This performance helps ensure a high degree of confidence in quantitation data obtained from a broad range of sample types.
• Highly sensitive quantitation using proven TaqMan real-time qPCR technology
• Offers consistent performance across the expected range of DNA fragment sizes
• Integrated system includes sample preparation kit, master mix, TaqMan primer/probe mix, and Vero genomic DNA standard
• Manual and automated sample preparation, optimized for quantitative recovery from complex sample matrices
This kit is available with or without sample preparation reagents in the form of a PrepSEQ Residual DNA Sample Preparation Kit, which is optimized for highly efficient residual DNA recovery from complex mixtures of proteins, buffers, and salts.
The resDNASEQ Quantitative Vero DNA Kit is designed to provide highly sensitive detection of Vero DNA, allowing you to use small sample volumes to help generate accurate results. The broad linear range of TaqMan real-time qPCR technology allows you to test samples with variable levels of Vero DNA in the same assay, such as in-process samples with higher amounts of DNA or bulk drug substance with very low amounts.
Easy to use, results in under 4 hours
The easy workflow of the resDNASEQ Quantitative Vero DNA Kit consists of sample preparation, assay setup, and instrument run and data analysis—all of which you can perform typically in under 4 hours. Start by extracting DNA from your in-process purification and drug substance samples. Then perform qPCR analysis to compare the amount of Vero DNA in your test samples to a standard curve generated with known amounts of purified Vero genomic DNA standard. The resDNASEQ system includes almost everything you need to carry out these steps, from sample preparation kit to master mix to TaqMan primer/probe mix to Vero genomic DNA standard, as well as step-by-step instructions.
Consistent performance even with fragmented DNA
For accurate quantitation of residual Vero DNA, assay results must be unaffected by the size of the DNA molecules present in the test sample. With the resDNASEQ system you get consistent assay performance across a range of DNA fragment sizes.
Specifications
Specifications
| Content And Storage | resDNASEQ Vero DNA Control • Vero DNA Control, 30 ng/μL, 40 μL, store at –15 to –25°C • DNA Dilution Buffer (DDB), 7 mL, store at –15 to –25°C before first use and 2−8°C after first use resDNASEQ Vero DNA Real-Time PCR Reagents • Environmental Master Mix (2X), 2 x 0.75 mL, store at –15 to –25°C before first use and 2−8°C after first use, protect from light • Vero DNA Assay Mix (10X), 300 μL, store at –15 to –25°C, protect from light • Negative Control (water), 1.0 mL, store at –15 to –25°C before first use and 2−8°C after first use PrepSEQ Nucleic Acid Extraction Kit • Lysis Buffer (2 bottles, 50 mL/bottle) • Binding Solution/Isopropanol (1 empty bottle) • Wash Buffer Concentrate (2 bottles, 26 mL/bottle) • Elution Buffer (1 bottle, 25 mL) • Proteinase K Buffer (1 bottle, 50 mL) • Magnetic Particles (2 tubes, 1.5 mL/tube) • Proteinase K (20 mg/mL) (1 tube, 1.25 mL), store at or below –20°C • PK Buffer II (1 bottle, 11 mL) Store all at room temperature unless otherwise indicated. PrepSEQ Residual DNA Sample Preparation Kit • Proteinase K (20 mg/mL), 1 tube, 1.25 mL • Yeast tRNA (10 mg/mL), 1 tube, 0.5 mL • Glycogen (5 mg/mL), 2 tubes, 1.0 mL/tube Store all at or below –20°C. |
| Detection Method | Primer-probe |
| Format | Kit |
| PCR Method | qPCR |
| Host Cell | Vero |
| Label or Dye | FAM |
| Product Line | resDNASEQ |
| Quantity | 100 reactions |
| No. of Reactions | 100 Reactions |
| Includes | Assay Kit and Sample Preparation |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.