missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Research Grade FBS, South America Origin

Research grade Fetal Bovine Serum of South America Origin. Certificate of analysis available.

Brand:  Cytiva SV30160.02

56.51 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.


Product Code. 10166413

  • £64.95 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Supplement research cell culture growth with South America-sourced quality HyClone Fetal Bovine Serum.
  • Sourced in EU-approved South American countries with complete traceability back to original source
  • Low in antibodies and high in growth factors
  • Produced using true pool technology for uniformity and consistency from bottle to bottle
  • Filtered through three sequential 100nm (0.1um) pore size-rated filters
  • Virus panel testing according to 9 CFR 113.53
TRUSTED_SUSTAINABILITY
Specifications

Specifications

South America
Sterile
Endotoxin, Hemoglobin
100 mL
Sequential Triple-filtered, 100 nm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers