missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Reserpine, Tocris Bioscience™
Click to view available options
Quantity:
1 g
Description
Binds the vesicular monoamine transporter (VMAT2) and inhibits transport of biogenic amines into adrenal chromaffin granules and synaptic vesicles. Causes depletion of biogenic amine stores. Antihypertensive and antipsychotic.
Specifications
Specifications
CAS | 50-55-5 |
Molecular Formula | C33H40N2O9 |
Synonym | reserpine, serpalan, serpasil, serpivite, apoplon, deserpine, elserpine, hiserpia, hypersil, raunervil |
InChI Key | QEVHRUUCFGRFIF-MDEJGZGSSA-N |
SMILES | COC1C(CC2CN3CCC4=C(C3CC2C1C(=O)OC)NC5=C4C=CC(=C5)OC)OC(=O)C6=CC(=C(C(=C6)OC)OC)OC |
IUPAC Name | methyl (1R,15S,17R,18R,19S,20S)-6,18-dimethoxy-17-(3,4,5-trimethoxybenzoyl)oxy-1,3,11,12,14,15,16,17,18,19,20,21-dodecahydroyohimban-19-carboxylate |
Molecular Weight (g/mol) | 608.688 |
PubChem CID | 5770 |
ChEBI | CHEBI:28487 |
Formula Weight | 608.68 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction