missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Reservoir Bottles for Automatic Burette

Spare 2000mL bottles for automatic burette manufactured of boro.3.3 glass.

Brand:  BRAND™ 23320

 View more versions of this product

Product Code. 10429641

  • £117.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Manufactured of boro 3.3 glass with a PP screw-cap and PE-LD pouring ring
  • Maximum operating temperature is 80°C (cleaning temperatures should not exceed 60°C)
  • Requires use of bottletop adapter - 33mm/STJ 29/32
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Amber
Automatic Burette
Reservoir Bottle
2000 mL
Borosilicate Glass
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.