missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Multi-Channel Reservoirs
Support full sample recovery with tapered reservoirs. Fisherbrand™ Multi-Channel Reservoirs are designed for superior handling with thicker, crush-resistant walls. Disposable reservoirs are available in two sizes of sterile or nonsterile options, plus a clear PVC option for better sample visibility.
£40.55 - £142.00
Specifications
Dimensions (L x W x H) | 145 x 85 x 25mm |
---|---|
Type | Reagent Reservoir |
Disposable | Single-use |
Product Code | Brand | Capacity (Metric) | Sterility | Material | Color | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Sterility | Material | Color | Price | Quantity & Availability | ||
11314035
|
Fisherbrand™
|
55 mL | Non-sterile | PVC | Transparent |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11344035
|
Fisherbrand™
|
100 mL | Non-sterile | Polystyrene | White |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11324035
|
Fisherbrand™
|
55 mL | Non-sterile | Polystyrene | White |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
15312630
|
Fisherbrand™
|
55 mL | Non-sterile | Polystyrene | White |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11334035
|
Fisherbrand™
|
55 mL | Sterile | Polystyrene | White |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11354035
|
Fisherbrand™
|
100 mL | Sterile | Polystyrene | White |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
- Thick walls of white polystyrene or clear PVC, with crush-resistant turrets at the corners
- Tapered reservoir bottoms and easy-pour spouts to aid full sample recovery
- Stackable, with tabs to prevent stacked reservoirs from sticking
- 55mL and 100mL volumes, both with 25mL graduation marks
- Sterile versions available
- Gamma-irradiated in an ISO-certified facility
- Individually sealed in Sani-Tear HDPE packets
Specifications
145 x 85 x 25mm | |
Reagent Reservoir | |
Single-use |