missing translation for 'onlineSavingsMsg'
Learn More

Residual Chlorine QC Sample, SPEX CertiPrep™

Residual Chlorine QC Sample, 2 mL, SPEX CertiPrep™. ISO Guide 34 Certified

Brand:  SPEX Certiprep 696

Product Code. 15608215

  • £124.00 / 2mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
This item is not returnable. View return policy

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

  • Residual Chlorine Sample
  • QC Sample
  • 2 mL vial yields up to 2L after dilution

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Residual Chlorine QC Sample
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Yields up to 2L after dilution
2 mL
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product intended for laboratory use only