missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Resiniferatoxin, 99+%, Thermo Scientific™
Click to view available options
Quantity:
1 mg
10 mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Resiniferatoxin, a potent capsaicin-like stimulator of peripheral nociceptors in the neonatal rat tail in vitro. Resiniferatoxin is used as an anti-inflammatory, anti-viral and anti-psoriatic uses. Diterpene ester related to phorbol but apparently not tumorigenic activates protein kinase C
Solubility
Soluble in DMSO to 100mM and to 50mM in ethanol
Notes
Resiniferatoxin (RTX) is susceptible to slow degradation if exposed to air. Even at -20°C, Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep in a dry place. Keep away from strong oxidizing agents. RTX must be stored in an inert atmosphere for long-term stability.
Resiniferatoxin, a potent capsaicin-like stimulator of peripheral nociceptors in the neonatal rat tail in vitro. Resiniferatoxin is used as an anti-inflammatory, anti-viral and anti-psoriatic uses. Diterpene ester related to phorbol but apparently not tumorigenic activates protein kinase C
Solubility
Soluble in DMSO to 100mM and to 50mM in ethanol
Notes
Resiniferatoxin (RTX) is susceptible to slow degradation if exposed to air. Even at -20°C, Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep in a dry place. Keep away from strong oxidizing agents. RTX must be stored in an inert atmosphere for long-term stability.
Specifications
Color | White |
Quantity | 10 mg |
UN Number | UN2811 |
Beilstein | 5371150 |
Solubility Information | Soluble in DMSO to 100mM and to 50mM in ethanol |
Formula Weight | 628.71 |
Percent Purity | ≥99% |
Physical Form | Solid |
Chemical Name or Material | Resiniferatoxin |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'