Learn More
Resorcinol, ACS, 99.0-100.5%, Thermo Scientific Chemicals
Description
Resorcinol is used in the synthesis of pharmaceuticals and various organic compounds. It is likewise used in plasticizers and diazo dyes. It can be applied in the qualitative determination of ketoses in the Seliwanoff test. Resorcinol-formaldehyde resin has shown useful application in endodontic therapy in russian red. It is an indispensable element for spectrophotometric determination of nitrate concentrations in seawater.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 109°C to 112°C |
Color | White to Cream |
Density | 1.272 g/mL |
pH | 4 to 6 |
Boiling Point | 280°C to 281°C |
Flash Point | 171°C (339°F) |
Assay Percent Range | 99.0 to 100.5% |
Odor | Phenol-like |
Quantity | 25 g |
UN Number | UN2876 |
Show More |
RUO – Research Use Only