missing translation for 'onlineSavingsMsg'
Learn More

Resorcinol, Certified AR for Analysis, Fisher Chemical™

£92.50 - £158.00

Chemical Identifiers

CAS 108-46-3
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.112
MDL Number 2269
InChI Key GHMLBKRAJCXXBS-UHFFFAOYSA-N
Synonym resorcinol, 1,3-benzenediol, resorcin, 1,3-dihydroxybenzene, 3-hydroxyphenol, m-hydroquinone, resorcine, m-dihydroxybenzene, m-hydroxyphenol, m-benzenediol
PubChem CID 5054
ChEBI CHEBI:27810
IUPAC Name benzene-1,3-diol
SMILES C1=CC(=CC(=C1)O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10529390
View Documents
Fisher Chemical
R/0200/48
100 g
£92.50
100g
Please to purchase this item. Need a web account? Register with us today!
10040350
View Documents Promotion Details
Fisher Chemical
R/0200/50
250 g
£158.00
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

108-46-3
110.112
GHMLBKRAJCXXBS-UHFFFAOYSA-N
5054
benzene-1,3-diol
C6H6O2
2269
resorcinol, 1,3-benzenediol, resorcin, 1,3-dihydroxybenzene, 3-hydroxyphenol, m-hydroquinone, resorcine, m-dihydroxybenzene, m-hydroxyphenol, m-benzenediol
CHEBI:27810
C1=CC(=CC(=C1)O)O

Specifications

108-46-3
Beige
281°C
C6H6O2
resorcinol, 1,3-benzenediol, resorcin, 1,3-dihydroxybenzene, 3-hydroxyphenol, m-hydroquinone, resorcine, m-dihydroxybenzene, m-hydroxyphenol, m-benzenediol
C1=CC(=CC(=C1)O)O
110.112
CHEBI:27810
Solid
109°C
4.4
Glass Jar
2269
GHMLBKRAJCXXBS-UHFFFAOYSA-N
benzene-1,3-diol
5054
110.11g/mol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Resorcinol

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications