missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Resorcinol, Extra Pure, SLR, Fisher Chemical™
Chemical Identifiers
CAS | 108-46-3 |
---|---|
Molecular Formula | C6H6O2 |
Molecular Weight (g/mol) | 110.112 |
MDL Number | 2269 |
InChI Key | GHMLBKRAJCXXBS-UHFFFAOYSA-N |
Synonym | resorcinol, 1,3-benzenediol, resorcin, 1,3-dihydroxybenzene, 3-hydroxyphenol, m-hydroquinone, resorcine, m-dihydroxybenzene, m-hydroxyphenol, m-benzenediol |
PubChem CID | 5054 |
ChEBI | CHEBI:27810 |
IUPAC Name | benzene-1,3-diol |
SMILES | C1=CC(=CC(=C1)O)O |
Description
Resorcinol, Extra Pure, SLR
Chemical Identifiers
108-46-3 | |
110.112 | |
GHMLBKRAJCXXBS-UHFFFAOYSA-N | |
5054 | |
benzene-1,3-diol |
C6H6O2 | |
2269 | |
resorcinol, 1,3-benzenediol, resorcin, 1,3-dihydroxybenzene, 3-hydroxyphenol, m-hydroquinone, resorcine, m-dihydroxybenzene, m-hydroxyphenol, m-benzenediol | |
CHEBI:27810 | |
C1=CC(=CC(=C1)O)O |
Specifications
108-46-3 | |
Beige | |
281°C | |
C6H6O2 | |
resorcinol, 1,3-benzenediol, resorcin, 1,3-dihydroxybenzene, 3-hydroxyphenol, m-hydroquinone, resorcine, m-dihydroxybenzene, m-hydroxyphenol, m-benzenediol | |
C1=CC(=CC(=C1)O)O | |
110.112 | |
CHEBI:27810 | |
Solid |
109°C | |
4.4 | |
Glass Jar | |
2269 | |
GHMLBKRAJCXXBS-UHFFFAOYSA-N | |
benzene-1,3-diol | |
5054 | |
110.11g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Resorcinol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.