missing translation for 'onlineSavingsMsg'
Learn More

Resorcinol-O,O'-diacetic acid, 97+%, Thermo Scientific™

Product Code. 11367465
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-39-6
Molecular Formula C10H10O6
Molecular Weight (g/mol) 226.184
MDL Number MFCD00016696
InChI Key ZVMAGJJPTALGQB-UHFFFAOYSA-N
Synonym resorcinol-o,o'-diacetic acid, m-phenylenedioxydi acetic acid, 2-3-carboxymethoxy phenoxy acetic acid, 2,2'-1,3-phenylenebis oxy diacetic acid, 2,2'-1,3-phenylenebis oxy bis-acetic acid, acetic acid,2,2'-1,3-phenylenebis oxy bis, 3-carboxymethoxy phenoxyacetic acid, m-phenylenedioxy diacetic acid, m-phenylenedioxydi
PubChem CID 66884
IUPAC Name 2-[3-(carboxymethoxy)phenoxy]acetic acid
SMILES C1=CC(=CC(=C1)OCC(=O)O)OCC(=O)O

Specifications

Melting Point 192°C to 196°C
Quantity 5 g
Beilstein 2135777
Formula Weight 226.19
Percent Purity ≥97%
Chemical Name or Material Resorcinol-O,O'-diacetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'