missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva RESOURCE™ Q IEX Columns
Use for intermediate lab-scale purification and large-scale polishing of biomolecules Cytiva Life Sciences™ RESOURCE™ Q IEX Columns are strong anion exchanger columns.
Brand: Cytiva 17-1177-01
Code : GE
Additional Details : Weight : 0.08500kg
Description
- Rigid, monodispersed, spherical particles with controlled pore-size distribution offer excellent flow characteristics, sample loading of up to 25mg/mL and stability in organic solvents and pH extremes
- Resource columns with 1mL bed volume have separation times of 3 minutes or less at 9.6mL/min. and approximately 20 min. using a peristaltic pump at 1mL/min.
Specifications
Dynamic Capacity: 40-54 mg BSA/ml | |
PEEK | |
>10 mL/min. | |
Quaternary ammonium | |
6.4 mm | |
15 μm | |
1 x 1 mL |
Connectors for high- and low-pressure chromatography equipment and instructions | |
Polystyrene/divinylbenzene | |
RESOURCE Q columns are prepacked with SOURCE 15Q, a strong anion exchanger for high resolution polishing purification of proteins at high flow rates. | |
4°C to 30°C in 20% Ethanol | |
1 mL | |
2 to 12 |