missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva RESOURCE™ S IEX Columns
RESOURCE prepacked columns for fast separation. Resource S IEX Columns are prepacked with Source 15S, a strong cation exchanger for intermediate lab-scale purification and large-scale polishing of biomolecules with ion exchange chromatography.
£721.00 - £2178.45
Specifications
Description | RESOURCE S |
---|---|
Kit Contents | Connectors for high- and low-pressure chromatography equipment and instructions |
Column Material | PEEK |
Matrix | Polystyrene/divinylbenzene |
For Use With (Application) | RESOURCE S columns are prepacked with SOURCE 15S, a strong cation exchanger for high-resolution polishing purification of proteins, at high flow rates. |
Product Code | Brand | Volume (Metric) | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Quantity | Price | Quantity & Availability | ||||
10695565
|
Cytiva
17-1178-01 |
1 mL | 1 x 1 mL |
£721.00
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11574975
|
Cytiva
17-1180-01 |
6 mL | 1 x 6 mL |
£2178.45
6mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Rigid, monodispersed, spherical particles with controlled pore-size distribution offer excellent flow characteristics, sample loading of up to 25mg/mL and stability in organic solvents and pH extremes
- Resource columns with 1mL bed volume have separation times of 3 minutes or less at 9.6mL/min. and approximately 20 min. using a peristaltic pump at 1mL/min.
Specifications
RESOURCE S | |
PEEK | |
RESOURCE S columns are prepacked with SOURCE 15S, a strong cation exchanger for high-resolution polishing purification of proteins, at high flow rates. | |
4°C to 30°C, 0.2M Sodium Acetate in 20% Ethanol | |
2 to 13 |
Connectors for high- and low-pressure chromatography equipment and instructions | |
Polystyrene/divinylbenzene | |
Sulphonate group | |
15 μm |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- resource S
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P235-Keep cool.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva RESOURCE™ S IEX Columns