missing translation for 'onlineSavingsMsg'
Learn More

3M™ 6000 Series Half Face Mask Respirator

Half-face mask respirators 6000 Series are economical, low-maintenance, and extremely lightweight. Probed reusable respirator assembly 6000Q series are used for quantitative fit purposes only.

Brand:  3M™ 7000146849

15.59 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 12652555

  • £31.15 / Each
Estimated Shipment: 15-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Combines features of both reusable and disposable respirators to provide effective protection. 3M™ Half Face Mask Respirator 6000 Series are economical, low-maintenance, simple-to-handle, and extremely lightweight.

  • Reusable half masks are economical, low-maintenance, simple-to-handle, and extremely lightweight
  • Used with 3M™ Supplied Air Systems
  • Bayonet Connection System allows connection to a broad range of twin lightweight filters to protect against gases, vapors, and particulates
  • Twin filter design provides lower breathing resistance
  • Probed reusable respirator assembly 6000 Series used for quantitative fit testing (17-987-3A & -3B)

Compliance

NIOSH approved under 42 CFR 84 standard for use against a variety of respiratory hazards when used as designed with appropriate cartridges and filters; in Canada, CSA standard Z94.4 and applicable local government requirements must be met for approval.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

EN 140:1998
Thermoplastic Elastomer
Large
6300L
Gray
Half-Mask Respirator
Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, painting, remediation, sanding, welding
4-point
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3M™ 6000 Series Half Face Mask Respirator > 6000 Series

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.