Learn More
Moldex™ Respirator Fit Test Kit
Allows employers to manage their own fit testing requirements
Brand:  Moldex™ 10301
Product Code. 12302309
  • £331.00 / Each
Currently Unavailable: We have no information when this product will be available
View alternative products
Explore more special offers
Includes: Hood (1), Nebulisers (2), 6 x 2.5mL Bitrex™ Sensitivity Solution ampoules, 6 x 2.5mL Bitrex™ Test Solution ampoules, Spare parts for nebulisers (2), Instruction CD (1)
Description
Description
  • Fit test kit to perform qualitative fit tests
  • Contains everything needed for multiple fit tests
  • Free instruction CD
  • Packaged in sturdy carry case
Specifications
Specifications
Fit Test Kit
Protection against dust, mite, and fume
1 Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Moldex™ Respirator Fit Test Kit >