missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Restek™ 1/16 in. Tubing Cutting Pliers
Click to view available options
Size (English) Tubing:
0.06 in.
Description
- Cut quickly, reducing distortion
- Cut clean, eliminating need for deburring
Specifications
Specifications
| Type | Tubing Cutting Pliers |
| Size (English) Tubing | 0.06 in. |
| Size (Metric) Tubing | 0.16 cm |
| For Use With (Application) | Ideal for Cutting 0.16 cm Tubing by Eliminating Tubing Distortion |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction