missing translation for 'onlineSavingsMsg'
Learn More

Restek™ 11mm GC Snap-Top Cap with Septum

An effecitve alternative when there is no need to crimp caps

Brand:  Restek™ 21734

Encompass_Preferred

 View more versions of this product

Product Code. 13119642

  • £528.77 / Pack of 1000
Estimated Shipment: 13-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Snaps on any crimp-style vial
  • Clear
  • Polypropylene

Specifications

Specifications

PTFE/Silicone
Polypropylene
11 mm
Clear
1000/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers