missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Restek™ 11mm GC Snap-Top Cap with Septum
An effecitve alternative when there is no need to crimp caps
Brand: Restek™ 21729
This item is not returnable.
View return policy
Description
- Snaps on any crimp-style vial
- Clear
- Polypropylene
Specifications
PTFE | |
Polypropylene | |
11 mm |
Clear | |
100/Pk. |