Learn More
Restek™ Capillary Column Caps
Attach to the column in seconds to form an airtight seal.
Brand: Restek™ 21044
This item is not returnable.
View return policy
Description
- Increase column lifetime by preventing moisture and air from entering the column during storage
- Made from glass
- Color-coded for identifying detector and injector ends
- Not recommended for reuse
Specifications
Column Caps |