Learn More
Restek™ Detector Lamps
Used for waters detectors or for Agilent HPLC Systems
Brand: Restek™ 25408
Code : Z3
Additional Details : Weight : 0.50000kg
Description
- Standard lamps have nominal 1,000-hour life
Specifications
1 Chromatography Detector Lamp | |
Deuterium Lamp | |
996, 2996 Water Detector |