missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Filter and Baseplate Kits

Single-position mounting baseplate with 0.125 in. inlet/outlet fittings (06-711-828)

Brand:  Restek™ 22019

Code : Z3

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 13171412

  • £601.37 / Each
Estimated Shipment: 27-08-2024
to see stock.

Description

Description

Carrier Gas Cleaning Kit

  • Single-position mounting baseplate with 0.125 in. inlet/outlet fittings (06-711-828)
  • Triple filter for oxygen, moisture, and hydrocarbon impurities (06-711-824)
Fuel Gas Purification Kit
  • Single-position mounting baseplate with 0.125 in. inlet/outlet fittings (06-711-828)
  • Fuel gas filter for moisture and hydrocarbon impurities (06-711-826)

Specifications

Specifications

Carrier Gas Cleaning Kit
Filtering oxygen, moisture, and hydrocarbon impurities
Mounting base plate, 0.12 in. inlet/outlet fittings, and oxygen/moisture/hydrocarbon triple filter
Filter and Baseplate Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers