missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Intermediate-Polarity Deactivated Fused Silica Guard Columns and Transfer Lines

Most common solvents easily wet and create a uniform film on the tubing surface

Brand:  Restek™ 10044

Encompass_Preferred

 View more versions of this product

Product Code. 12935199

  • £94.38 / Each
Estimated Shipment: 07-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Wide range of applications
  • Maximum temperature 325°C
  • Diameters greater than 0.10mm tested with Grob test mix to ensure high inertness

Specifications

Specifications

0.45 ± 0.04 mm
0.32 mm
1 Ea.
Intermediate-Polarity, Deactivated Fused Silica
5 m
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers