Learn More
Restek™ Intermediate-Polarity Deactivated Fused Silica Guard Columns and Transfer Lines
Most common solvents easily wet and create a uniform film on the tubing surface
POA - £804.65
Products 13
Product Code Brand Diameter (Metric) Inner Length (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Inner Length (Metric) Price Quantity & Availability  
12935189
Encompass_Preferred
View Documents Promotion Details
Restek™
10040
0.05 mm 5 m
£143.99
Each
View alternative products
Save Up to 
»
Estimated Shipment: 07-11-2024
to see stock.
Add to basket
10017611
Encompass_Preferred
View Documents Promotion Details
Restek™
10041
0.1 mm 5 m
£160.93
Each
View alternative products
Save Up to 
»
Estimated Shipment: 07-11-2024
to see stock.
Add to basket
12955189
Encompass_Preferred
View Documents Promotion Details
Restek™
10041
0.1 mm 5 m
£160.93
Each
View alternative products
Save Up to 
»
Estimated Shipment: 07-11-2024
to see stock.
Add to basket
12995189
Encompass_Preferred
View Documents Promotion Details
Restek™
10043
0.25 mm 5 m
£84.70
Each
View alternative products
Save Up to 
»
Estimated Shipment: 07-11-2024
to see stock.
Add to basket
10029429
Encompass_Preferred
View Documents Promotion Details
Restek™
10043
0.25 mm 5 m
£84.70
Each
View alternative products
Save Up to 
»
Estimated Shipment: 07-11-2024
to see stock.
Add to basket
12975189
Encompass_Preferred
View Documents Promotion Details
Restek™
10042
0.15 mm 5 m
£160.93
Each
View alternative products
Save Up to 
»
Estimated Shipment: 07-11-2024
to see stock.
Add to basket
12935199
Encompass_Preferred
View Documents Promotion Details
Restek™
10044
0.32 mm 5 m
£94.38
Each
View alternative products
Save Up to 
»
Estimated Shipment: 07-11-2024
to see stock.
Add to basket
12965199
Encompass_Preferred
View Documents Promotion Details
Restek™
10045
0.53 mm 5 m
£121.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-11-2024
to see stock.
Add to basket
12975199
Encompass_Preferred
View Documents Promotion Details
Restek™
10045-050
0.53 mm 5 m This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12915199
Encompass_Preferred
View Documents Promotion Details
Restek™
10043-600
0.25 mm 5 m
£423.50
Pack of 6
View alternative products
Save Up to 
»
Estimated Shipment: 07-11-2024
to see stock.
Add to basket
12985189
Encompass_Preferred
View Documents Promotion Details
Restek™
10042-600
0.15 mm 5 m
£804.65
Pack of 6
View alternative products
Save Up to 
»
Estimated Shipment: 07-11-2024
to see stock.
Add to basket
12995199
Encompass_Preferred
View Documents Promotion Details
Restek™
10045-600
0.53 mm 5 m
£605.00
Pack of 6
View alternative products
Save Up to 
»
Estimated Shipment: 07-11-2024
to see stock.
Add to basket
12925209
Encompass_Preferred
View Documents Promotion Details
Restek™
10046-600
0.18 mm 5 m
£550.55
Pack of 6
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 07-11-2024
to see stock.
Add to basket
Description
Description
  • Wide range of applications
  • Maximum temperature 325°C
  • Diameters greater than 0.10mm tested with Grob test mix to ensure high inertness