Learn More
Restek™ MicroPulse Pulse Dampers
Compact unit can be placed almost anywhere
£860.31
Specifications
Type | Minipulse Pulse Dampener |
---|---|
For Use With (Application) | For Applications Where Minimizing The Total System Volume is Critical |
Volume (Metric) | 160 μL |
Description
The MiniPulse pulse damper is ideal for applications where minimizing the total system volume is critical.
- Compact unit
- Small
- 160μL dead volume at atmospheric pressure
- Diameter: 2.5 x 1.5 in.
Specifications
Minipulse Pulse Dampener | |
For Applications Where Minimizing The Total System Volume is Critical | |
160 μL |