Learn More
Restek™ Rxi™-XLB GC Columns
General-purpose columns exhibiting extremely low bleed
Brand:  Restek™ 13726
Encompass_Preferred
 View more versions of this product
Product Code. 10610447
  • £1360.04 / Each
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Especially well-suited for analysis of active, high molecular weight compounds with sensitive GC/MS systems including ion trap detectors
  • Temperature range 30° to 360°C for 15m and 30m lengths: longer columns may have a slightly reduced maximum temperature
  • Well-suited for U.S. EPA Method 525 for analysis of semivolatile compounds in drinking water
Pesticides, PCB cogeners
Specifications
Specifications
0.25 mm
Pesticide Analysis, PCB Congener Analysis, PAHs Analysis
30°C to 340/360°C
60 m
Fused Silica
Rxi™-XLB
0.25 μm
0.25 μm
Low polar