Learn More
Restek™ Septum Nuts for use with Agilent GCs
For use with Agilent 5890, 6850, 6890, 7890 split/splitless injectors.
Brand:  Restek™ 20631
Encompass_Preferred
 View more versions of this product
Product Code. 12902569
  • £60.50 / Each
Estimated Shipment: 29-10-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Needle guide allows easy penetration and prevents septum coring
  • Made of high-quality stainless steel
  • Manual injection septum nut allows use of 26-gauge needles for on-column injections
  • Autosampler and PTV septum nut allows use of 23-gauge needles
Specifications
Specifications
Septum Nut
Manual Injection (for 26-Gauge Needles)