Learn More
Restek™ Stabilwax™-DA Capillary Columns
Similar in polarity to DB-FFAP, OV-351, NUKOL FFAP, HP FFAP
Brand:  Restek™ 11065
Encompass_Preferred
 View more versions of this product
Product Code. 12975689
  • £894.19 / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Polyethylene glycol stationary phase has high thermal stability up to 250°C (482°F)
  • Crossbond™ Carbowax™ technology provides low bleed, increased column life, and solvent rinsability
  • Resist oxidation and thermal breakdown
  • Allow free fatty acids from C1 to C20 to be analyzed without derivatization; also works well for some inorganic acids
  • Individual test chromatogram is provided with each column
Applications:
Underivatized free fatty acids
Specifications
Specifications
Acidic
Stabilwax™-DA
Free (underivatized) Acids, Some Inorganic Acids Analysis
40°C to 230/240°C
30 m
Polar
0.53 mm
G25, G35
1.5 μm
1.5 μm
Stainless Steel
Fused Silica