Learn More
Restek™ Stainless Steel Tube Brushes/Surface Brush Set
Click to view available options
Packaging Quantity:
Set of 4
Description
- Use surface brush to remove residue that builds up on metal detector jets and electronic contacts
- Set of four brushes - three tube brushes (3/8 in., 3/16 in., and 1/4 in.) and one surface brush
Specifications
Specifications
| Type | Tube Brushes/Surface Brushes |
| For Use With (Application) | Remove Residue That Builds up on Metal Detector Jets and Electronic Contacts and for Cleaning Dirty Collectors, Injector Ports, and Detector Ports. |
| Packaging Quantity | Set of 4 |
| Material | Stainless Steel |