missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Restek™ Straight Inlet Liners for Splitless Injection
Used for samples containing a narrow molecular weight distribution and for those not prone to thermal decomposition
POA - £173.03
Specifications
Length (Metric) | 7.85 cm |
---|---|
Product Type Specs | IP Splitless Straight Inlet Liner |
For Use With (Equipment) | Agilent GCs Equipped with Split/Splitless Inlets |
Product Code | Brand | Diameter (Metric) Inner | Quantity | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Quantity | Includes | Price | Quantity & Availability | |||
12912649
|
Restek™
20712 |
0.2 cm | Each | Glass Splitless | |||||
12952739
|
Restek™
20772 |
0.4 cm | Each | Glass Splitless | |||||
13134592
|
Restek™
22400 |
0.4 cm | Each | Glass Splitless with Wool | |||||
13164592
|
Restek™
22401 |
0.4 cm | 5/Pk. | Glass Splitless with Wool |
£89.54
Pack of 5 |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||
10058314
|
Restek™
20713 |
0.2 cm | 5/Pk. | Glass Splitless |
£119.79
Pack of 5 |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||
12972649
|
Restek™
20713 |
0.2 cm | 5/Pk. | Glass Splitless | |||||
12932659
|
Restek™
20714 |
0.2 cm | 25/Pk. | Glass Splitless | |||||
12922749
|
Restek™
20773 |
0.4 cm | 5/Pk. | Glass Splitless |
£96.80
Pack of 5 |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||
10097781
|
Restek™
20773 |
0.4 cm | 5/Pk. | Glass Splitless |
£96.80
Pack of 5 |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||
12932749
|
Restek™
20773-202.5 |
4.0 mm | 5/Pk. | Splitless/HP w/Glass Wool | |||||
12912759
|
Restek™
20774 |
0.4 cm | 25/Pk. | Glass Splitless | |||||
13114602
|
Restek™
22402 |
0.4 cm | 25/Pk. | Glass Splitless with Wool | |||||
13184602
|
Restek™
22404 |
0.4 cm | 5/Pk. | Quartz Splitless with Wool |
£173.03
Pack of 5 |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||
Description
- Useful for splitless mode of operation and slow injections, small (<2μL) sample volumes, and high-boiling point solvent systems
- Compatible with Agilent™/Finnigan™ columns
- Liners should be packed with wool or fused silica beads to trap nonvolatile residue and prevent column contamination when analyzing dirty samples; also, packing in wool is recommended to aid in vaporization of high molecular weight compounds and to minimize discrimination
- O.D. × Length: 6.5 × 78.5mm
Specifications
7.85 cm | |
IP Splitless Straight Inlet Liner | |
Agilent GCs Equipped with Split/Splitless Inlets |