Learn More
Thermo Scientific™ Restore™ Fluorescent Western Blot Stripping Buffer
Quickly and gently remove primary and near-infrared (IR) dye-labeled secondary antibodies from Western blots.
£55.50 - £173.00
Specifications
Form | Liquid |
---|---|
Product Line | Restore™ |
Content And Storage | Upon receipt store at room temperature. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11835814
|
Thermo Scientific™
62299 |
20 mL |
£55.50
20mL |
Estimated Shipment: 09-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11577900
|
Thermo Scientific™
62300 |
100 mL |
£173.00
100mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Thermo Scientific™ Restore Fluorescent Western Blot Stripping Buffer is a gentle and highly effective reagent for quickly removing primary and near-infrared (IR) dye-labeled secondary antibodies from Western blots.
Restore Fluorescent Western Blot Stripping Buffer enables the reuse of PVDF membranes, simplifying the Western blot optimization process and allowing the same blot to be reprobed with different primary antibodies to detect alternative targets. Restore Fluorescent Western Blot Stripping Buffer is for use with low-fluorescence PVDF membrane only (Part No. 22860).Highlights:
Fast – strip blots in only 15 minutes at room temperature
Saves time – no need to run new gels and prepare a new blot
Conserve samples – reprobe the same PVDF membrane for multiple targets
Economical – less expensive than other commercially available stripping buffers
Efficient – effectively strips blots the first time
Specifications
Liquid | |
Restore™ | |
Upon receipt store at room temperature. |
For Research Use Only. Not for use in diagnostic procedures.