missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Restore™ PLUS Western Blot Stripping Buffer

Remove high-affinity antibodies from membranes so they can be reprobed and detected with chemiluminescent substrates.

Brand:  Thermo Scientific™ 46428


 View more versions of this product

Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11829380

  • £56.75 / 30mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Restore PLUS Western Blot Stripping Buffer is an advanced formula for removing bound primary and secondary antibodies from membranes so they can be reprobed and detected with chemiluminescent substrates.

Restore PLUS Buffer is an alternative formulation of the original Thermo Scientific Restore Western Blot Stripping Buffer. Restore PLUS Stripping Buffer was designed for use with antibodies that are difficult to remove from Western blots and require longer incubation times or incubation temperatures greater than 22°C with gentler formulations. High-affinity antibodies can be quickly and effectively stripped from Western blots at room temperature without removing transferred proteins, thereby allowing multiple reprobes of the target.

Highlights:

  • Ready and easy to use – no dilution necessary; no offensive odors; store at room temperature

  • Compatible – use on nitrocellulose and PVDF membranes, whether still wet or already dry; works with practically any blocking buffer, enzyme conjugate and chemiluminescent substrate

  • Cost effective – save valuable time and samples; strip blots effectively the first time

  • Robust yet gentle – transferred proteins remain viable; strip the same blot up to five times

  • Flexible – strip and reprobe to optimize antibody concentrations or to detect a new antigen with different antibodies

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Western Blot Stripping Buffer
Liquid
Nitrocellulose, PVDF
30 mL
Chemiluminescence
Western Blot
Restore™
Upon receipt store at room temperature. Product shipped at ambient temperature.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • RESTORE PLUS WESTERN BLOT

Signal Word
  • Warning

Hazard Category
  • Substances and mixtures corrosive to metals Category 1

Hazard Statement
  • H290-May be corrosive to metals.

Precautionary Statement
  • P234-Keep only in original container.

Supplemental information
  • MIXTURE LIST-Contains : Anionic Surfactant ANIONIC SURFACT ( 1-3)

SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZL400224 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number46428
Lot NumberZE392456 Certificate TypeCertificate of Analysis Date07/06/2024 Catalog Number46428
Lot NumberYF372189 Certificate TypeCertificate of Analysis Date11/07/2023 Catalog Number46428
Lot NumberYA364944 Certificate TypeCertificate of Analysis Date11/02/2023 Catalog Number46428
Lot Numberspecs Certificate TypeCertificate of Analysis Date11/02/2023 Catalog Number46428
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Restore™ PLUS Western Blot Stripping Buffer > Stripping buffer; 30mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.