Learn More
Thermo Scientific™ Restore™ Western Blot Stripping Buffer
Safely and effectively remove primary and secondary antibodies from nitrocellulose and PVDF membranes to allow chemiluminescent Western blots to be reprobed.
£53.75 - £854.00
Specifications
Form | Liquid |
---|---|
Product Line | Restore™ |
Content And Storage | Storage: Upon receipt store at room temperature or 4°C. Product shipped at ambient temperature. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10485315
|
Thermo Scientific™
21062 |
30 mL |
£53.75
30mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10057103
|
Thermo Scientific™
21059 |
500 mL |
£198.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11874524
|
Thermo Scientific™
21063 |
5 L |
£854.00
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thermo Scientific™ Restore Western Blot Stripping Buffer safely and effectively removes primary and secondary antibodies from nitrocellulose and PVDF membranes to allow chemiluminescent Western blots to be reprobed.
Performing gel electrophoresis and duplicate immunoblot assays to test new primary antibodies or antibody concentrations is time-consuming and expensive. Restore Western Blot Stripping Buffer eliminates this waste when detecting immunoblots with chemiluminescent Western blotting substrates. The reagent allows clean and efficient removal of primary and secondary antibodies from immunoblots without removing or damaging the immobilized antigen. This allows blots to be stripped and reprobed with confidence.
Highlights:
Saves time – no need to re-run gels and blots
Saves costly sample – re-probe the membrane using the same target sample
Effective – formulation is more efficient at stripping antibodies than homemade buffers
Gentle – does not damage the target antigen during stripping allowing efficient reprobing
Odor-free – no mercaptans means no acrid odors
Economical – less expensive than other commercial stripping buffers
Reuse a nitrocellulose or PVDF blot to detect a different target with a different primary antibody; Reprobe a blot to correct or optimize antibody concentrations that were ineffective the first time.
Specifications
Liquid | |
Restore™ | |
Storage: Upon receipt store at room temperature or 4°C. Product shipped at ambient temperature. |
Safety and Handling
- Restore Western Blot Stripping Buffer
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P234-Keep only in original container.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P406-Store in corrosive resistant/ container with a resistant inner liner.
Supplemental information
- MIXTURE LIST-Contains : Organo Phosphine
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.