missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ret Antibody (132507) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB718JF585
This item is not returnable.
View return policy
Description
Ret Monoclonal antibody specifically detects Ret in Human samples. It is validated for Western Blot,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| Ret | |
| Monoclonal | |
| Janelia Fluor 585 | |
| CDHF12MTC1, CDHR16, EC 2.7.10, Hirschsprung disease 1, Hirschsprung disease), MEN2A, MEN2B, multiple endocrine neoplasia and medullary thyroid carcinoma 1, ret proto-oncogene, RET transforming sequence | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Ret/Fc Chimera, Leu29-Arg635, Accession # P07949 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 132507 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cancer, Oncogenes, Phospho Specific, Protein Kinase, Protein Phosphatase, Signal Transduction, Tumor Suppressors | |
| 5979 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction