Learn More
Retinoic acid, 97%, Thermo Scientific Chemicals
All-trans-retinol, Vitamin A, retinol, CAS # 68-26-8, is a fat-soluble vitamin that binds retinoid receptors within cells.
£30.10 - £331.00
Chemical Identifiers
CAS | 302-79-4 |
---|---|
Molecular Formula | C20H28O2 |
Molecular Weight (g/mol) | 300.44 |
MDL Number | MFCD00001551 |
InChI Key | SHGAZHPCJJPHSC-YCNIQYBTSA-N |
Synonym | retinoic acid, tretinoin, vitamin a acid, all-trans-retinoic acid, trans-retinoic acid, renova, airol, retin-a, vesanoid, atra |
PubChem CID | 444795 |
ChEBI | CHEBI:15367 |
SMILES | C\C(\C=C\C1=C(C)CCCC1(C)C)=C/C=C/C(/C)=C/C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11485290
|
Thermo Scientific Alfa Aesar
044540.77 |
0.1 g |
£30.10
0.10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11465290
|
Thermo Scientific Alfa Aesar
044540.02 |
0.5 g |
£105.00
0.50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11475290
|
Thermo Scientific Alfa Aesar
044540.04 |
2 g |
£331.00
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Retinoic acid is used in dermatology drugs. It is used in the treatment of psoriasis, ichthyosis, follicular keratosis, acne, lichen planus, verrucous epidermal nevus and lichen psoriasis. It is also employed in promoting the proliferation and differentiation of epithelial cells. In addition to this, it exhibits some preventive effect on skin cancer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
• All-trans-retinol, commonly known as Vitamin A, is a fat-soluble essential nutrient
• All-trans-retinol is involved in hormonal signaling via the retinoid receptors
• It is an important regulator of cell division and apoptosis
• Deficiency can lead to vision, bone, immune, and skin disorders
Applications
• Suitable for research on cell differentiation, embryonic development, immunology and more
• Laboratory grade quality, for research use only
Chemical Identifiers
302-79-4 | |
300.44 | |
SHGAZHPCJJPHSC-YCNIQYBTSA-N | |
444795 | |
C\C(\C=C\C1=C(C)CCCC1(C)C)=C/C=C/C(/C)=C/C(O)=O |
C20H28O2 | |
MFCD00001551 | |
retinoic acid, tretinoin, vitamin a acid, all-trans-retinoic acid, trans-retinoic acid, renova, airol, retin-a, vesanoid, atra | |
CHEBI:15367 |
Specifications
302-79-4 | |
C20H28O2 | |
2057223 | |
14,8165 | |
Soluble in ethanol,acetate,dimethyl formamide,dimethyl sulfoxide,chloroform,methanol,acetone and dichloromethane. Insoluble in water and ether. | |
C\C(\C=C\C1=C(C)CCCC1(C)C)=C/C=C/C(/C)=C/C(O)=O | |
300.44 | |
CHEBI:15367 | |
98% | |
Retinoic acid |
180°C to 182°C | |
MFCD00001551 | |
Light sensitive | |
retinoic acid, tretinoin, vitamin a acid, all-trans-retinoic acid, trans-retinoic acid, renova, airol, retin-a, vesanoid, atra | |
SHGAZHPCJJPHSC-YCNIQYBTSA-N | |
(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid | |
444795 | |
300.44 | |
Crystalline |
Safety and Handling
GHS H Statement
H361-H302
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
P201-P202-P261-P264b-P270-P271-P280-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
H302+H312-H315-H319-H335-H361
missing translation for 'einecsNumber' : 206-129-0
missing translation for 'rtecsNumber' : VH6475000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Retinoic acid
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H361d-Suspected of damaging the unborn child.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : all-trans-Retinoic acid
RUO – Research Use Only