Learn More
R&L Enterprises™ Retort Stand Clamps
Enables articles from 2 to 90mm in diameter to be clamped securely. R&L Enterprises™ Retort Stand Clamps feature aluminum alloy jaws with other parts of plated brass.
Brand:  R&L Enterprises™ 9505/P
Code : 02
Additional Details : Weight : 0.78750kg
 View more versions of this product
Product Code. 11888203
  • £211.00 / Pack of 5
Estimated Shipment: 05-09-2024
to see stock.
Add to basket
Description
Description
Enables articles from 2 to 90mm in diameter to be clamped securely. R&L Enterprises™ Retort Stand Clamps feature aluminum alloy jaws with other parts of plated brass.
  • Design ensures a firm grip at all angles of opening
  • Cork liners for the jaws are well secured
  • Available with either metal or plastic clamping screw head
Specifications
Specifications
5/Pk.
Aluminum Alloy, Brass
For use with 2 to 90mm diameter
Clamp Retort Stand
Plastic headed clamping screw
Cork Lined