missing translation for 'onlineSavingsMsg'
Learn More

RETSCH PM 100 Planterary Ball Mill Grinding Balls

For use with Retsch PM 100 and PM 200 Planetary Ball Mills

Brand:  RETSCH 05.368.0094

44.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10289201

  • £76.15 / Each
Estimated Shipment: 07-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • In assorted diameters and materials
  • Smaller balls offered for colloidal milling

Specifications

Specifications

Grinding Ball
1 cm
PM 100 and PM 200 planetary ball mill
0.39 in.
Ideal for standard grinding
Zirconium Oxide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
RETSCH PM 100 Planterary Ball Mill Grinding Balls > Zirconium oxide; For standard grinding; 10mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.