missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RETSCH Safe-Lock Reaction Vials
Mixer mill accessories: reaction vials
Brand: RETSCH 22.749.0001
This item is not returnable.
View return policy
Description
- Capacity: 2mL
- Sold 1000/Pk.
Specifications
2 mL | |
0.54 dr. |