missing translation for 'onlineSavingsMsg'
Learn More

Retsch Retsch Test sieve 200x50mm 53µm ISO 3310/1

Brand:  Retsch 60.131.000053

Retsch Test sieve 200x50mm 53µm ISO 3310/1

Product Code. 15670269

  • ONLINE EXCLUSIVE PRICE
    £141.00
    £135.00 / Each
    Save £6.00
    4% Off
Only null left
Estimated Shipment: 18-03-2026
Add to basket
This item is not returnable. View return policy

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.