missing translation for 'onlineSavingsMsg'
Learn More

3M™ 6000 Series Reusable Full Face Mask Respirators

Product Code. 10238294
Click to view available options
Size:
Small
Medium
Large
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10238294

Brand: 3M™ 7100015052

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Offers lightweight comfort and ease of use. 3M™ 6000 Series Reusable Full Face Mask Respirators have the 3M Bayonet Connection System, enabling connection to a broad range of twin lightweight filters to protect against gases, vapours and particulates.

  • Large lens for wide field of view and excellent visibility
  • Well-balanced design with silicone face seal for greater comfort and durability
  • Unique center adapter directs exhaled breath and moisture downward, reducing valve debris and facilitating quick and easy cleaning
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 136:1998 (Class 1)
Connection Type Bayonet
Color Dark Gray
Material Silicone, Thermoplastic Elastomer
Product Type Reusable Full Face Mask Respirator
Size Large
Facepiece Type Full
For Use With (Application) For construction applications
For Use With (Equipment) 200 series filters and 600 series cartridges
Harness Style 4-point
Includes Bayonet connection, double gasket, phonic membrane, four-point head strap
Lens Type Polycarbonate Visor
Material (Lens) Polycarbonate
Quantity 1 Ea.
Weight (Metric) 400 g
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.