missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Reusable Half Face Mask and A2P3 Combination Filter Kit
This handy, ready-to-use kit includes a 3M™ Reusable Half Face Mask and an A2P3 filter, all in a reusable storage box.
£865.00
Specifications
Product Type | Ready-to-Use Respirator Pack |
---|---|
Includes | 3M™ Reusable Half Face Mask and an A2P3 Filter |
Quantity | 10/Cs. |
Product Code | Brand | Size | Model | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Model | Price | Quantity & Availability | ||||
17706993
|
3M™
7100265072 |
Small | HF-3014 |
£865.00
Pack of 10 |
Estimated Shipment: 09-09-2024
Log in to see stock. |
||||
17716993
|
3M™
7100265073 |
Medium | HF-3024 |
£865.00
Pack of 10 |
Estimated Shipment: 09-09-2024
Log in to see stock. |
||||
17776983
|
3M™
7100265044 |
Large | HF-3034 |
£865.00
Pack of 10 |
Estimated Shipment: 09-09-2024
Log in to see stock. |
||||
Descripción
- Includes 1 x 3M™ Reusable Half Face Mask and 1 x 3M™ Combination Filter CF32 A2P3 R D DT-4041E
- Offers A2P3 protection from certain organic vapors and particulate hazards
- Comes in a useful, reusable storage box
Especificaciones
Ready-to-Use Respirator Pack | |
3M™ Reusable Half Face Mask and an A2P3 Filter | |
10/Cs. |