Learn More
3M™ Reusable Half Face Mask and P3 Particulate Filter Kit
This handy, ready-to-use kit includes a 3M™ Reusable Half Face Mask, a P3 filter and a pre-filter, all in a reusable storage box.
Brand: 3M™ 7100265071
342.00 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 5.18300kg
Descripción
- Includes 1 x 3M™ Reusable Half Face Mask, 1 x 3M™ Particulate Filter PF10 P3 R D DT-1135E, 1 x 3M™ Prefilter SC-6600 and 1 x 3M™ Prefilter Holder SC-6602
- Offers P3 particulate protection
- Comes in a useful, reusable storage box
Especificaciones
Ready-to-Use Respirator Pack | |
3M™ Reusable Half Face Mask, P3 Filter and Pre-Filter | |
10/Cs. |
Large | |
HF-3031 |