Learn More
3M™ Reusable Half Face Mask Respirator
Product Code. 17746993
Click to view available options
Size:
Small
Medium
Large
Model:
HF-301
HF-302
HF-303
Unit Size:
Pack of 10
Product Code. 17746993
Brand: 3M™ 7100265229
Only null left
Add to basket
Add to basket
3M™ Reusable Half Mask has been designed with user comfort and convenience in mind. Compatible with DIN threaded, 3M™ Filters DT-Series, the HF-301 offers protection against a wide range of respiratory hazards.
  • Hybrid reflex seal contours and flexes to the user's face to aid wearer comfort
  • Seal check mechanism - when pressed during exhale, the mask inflates slightly showing that a good seal has been achieved
  • Integrated voice panel for enhanced voice intelligibility while wearing the mask
  • Nose pinch allows for improved integration with eyewear
  • Comfortable, durable, low profile head cradle & thick elasticated straps with simple rear fastening
TRUSTED_SUSTAINABILITY
Specifications
Product Type Half Facepiece Respirator
Size Large
For Use With (Equipment) DIN Threaded, 3M™ Filters DT-Series
Model HF-303
Quantity 10/Cs.