Learn More
3M™ Reusable Half Mask Respirator Kit A2P3 R
3M™ Reusable Half Mask Respirator Kit A2P3 R is a safe solution for common applications in industries such as agriculture, construction, and painting.
£70.24
Specifications
Certifications/Compliance | CE Approved |
---|---|
Filter Type | A2P3 R |
Product Type | Half Mask Respirator Kit |
Includes | Half mask and filter |
Description
- Ready to use respiratory kit with 3M™ Reusable Half Face Mask 7503L or 7503M and 3M™ Filters offering A2P3 R protection
- Half mask with a soft silicone face piece for comfort
- Bayonet connection fitting system to easily connect filters to mask
- 3M™ Cool Flow™ Valve provides easier breathing and reduced heat and moisture build-up
- Low profile, twin filter design half mask for a balanced secure fit
Specifications
CE Approved | |
A2P3 R | |
Half Mask Respirator Kit | |
Half mask and filter |