Learn More
3M™ Reusable Low Maintenance Half Face Mask Respirator
Click to view available options
Size:
Medium
Large
Unit Size:
Pack of 8
Beschreibung
- For economical, low-maintenance reusable masks that are easy to handle, use 3M™ Reusable Half Face Mask 6000 Series.
- Our reusable half masks are designed for simplicity, with an elastomeric material that’s lightweight and comfortable to wear over long shifts, a cradle head harness and easy-fasten neck strap.
- They provide protection against gases, vapours and particulates, and feature a bayonet-style connection that fits to a broad range of twin, lightweight filters, depending on your individual needs.
- The twin-filter design evenly balances weight across the mask, and the low profile means minimal impact to your field of vision and better compatibility with welding and grinding shields.
- They can be used with 3M™ Supplied Air Systems for your increased convenience and flexibility, and connect with all 3M™ 2000, 5000 and 6000 Series filters (except 3M™ 6098 or 6099).
- The 6000 Series has a specially designed range of spares and accessories to help you maintain the right level of comfort and protection when parts become worn or damaged.
- They’re available in three sizes; small, medium and large.
Spezifikation
Spezifikation
| Color | Gray |
| Design Type | Bayonet Style |
| Disposable | Reusable |
| Filter Type | Combination |
| Material | Thermoplastic Elastomer |
| Product Type | Reusable Half Facepiece |
| Size | Medium |
| With Exhalation Valve | No |
| Latex Free | Yes |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur