Learn More
3M™ Reusable Respirator Full Facepiece Faceshield Cover 6800
Reusable Respirator Face Shield & Lens Covers is our range of covers designed to extend the service life of 3M™ Reusable Respirator lenses.
Brand: 3M™ 7100188168
110.00 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 0.18140kg
Description
- Designed for use with 3M™ Reusable Respirators
- Helps to extend the service life of the mask when in use and when in storage
- Spare or replacement part
Specifications
Reusable Respirator Parts & Accessories | |
3M™ Reusable Respirator lenses |