missing translation for 'onlineSavingsMsg'
Learn More
Learn More
REV1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-09940-100UL
This item is not returnable.
View return policy
Description
REV1 Polyclonal specifically detects REV1 in Human samples. It is validated for Western Blot.Specifications
REV1 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
AIBP80, Alpha integrin-binding protein 80, DNA repair protein REV1, EC 2.7.7.-, FLJ21523, MGC163283, MGC26225, REV1 (yeast homolog)- like, REV1 homolog (S. cerevisiae), REV1- like, REV1L, REV1-like (yeast), Rev1-like terminal deoxycytidyl transferase | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human REV1 (XP_005264024). Peptide sequence INLIALPAFSQVDPEVFAALPAELQRELKAAYDQRQRQGENSTHQQSASA | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
51455 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction