Learn More
Grant Instruments™ Reversible Raised Shelf for Unstirred Water Baths
Allows two shelf depths to accommodates shallow vessels. Grant Instruments™ Reversible Raised Shelf for Unstirred Water Baths can be used with a variety of other Grant Heating Circulators and Heated Circulating Baths.
£130.38 - £239.56
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
11456407
|
Grant Instruments™
RS18H |
Half the area of SBB Aqua 18 Plus or Sub Aqua 18 Pro |
£174.90
Each |
In Stock
Log in to see stock. |
|||||
11466407
|
Grant Instruments™
RS28H |
Half the area of SBB Aqua 26 Plus or Sub Aqua 26 Pro |
£184.44
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||||
11446407
|
Grant Instruments™
RS14H |
Half the area of SBB Aqua 12 Plus, Sub Aqua 12 Pro, or Sub Aqua Pro Dual |
£130.38
Each |
In Stock
Log in to see stock. |
|||||
11476407
|
Grant Instruments™
RS36H |
Half the area of SBB Aqua 34 Plus or Sub Aqua 34 Pro |
£239.56
Each |
In Stock
Log in to see stock. |
|||||
Description
- Choose from a variety of sizes to fit your tank.
- For use with either Stainless Steel and Plastic tank models
Specifications
Stainless Steel |