Learn More
Labconco™ ReVo MSC IV Bar
Bar supports intravenous solution bottles and bags. Kit includes IV bar and mounting hardware.
Brand: Labconco™ R3858601
This item is not returnable.
View return policy
Description
- IV Bar and mounting hardware compatible with various biosafety cabinets, including Purifier Axiom Biosafety Cabinets, Purifier Cell Logic and Cell Logic+ Biosafety Cabinets, Purifier Logic and Logic+ Biosafety Cabinets, Purifier Logic and PuriCare Procedure Stations and ReVo Microbiological Safety Cabinets.
Specifications
IV Bag Holder Kit | |
1200 x 20 x 20 mm | |
2.0 kg |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
Labconco™ ReVo MSC IV Bar > Length: 1200 mm
Spot an opportunity for improvement?Share a Content Correction